Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acap2
Entrez Gene ID
78618
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 2
Gene Aliases
4832442G16, 9530039J15Rik, CNT-B2, Centb2, mKIAA0041
Chromosome Location
Chr.16: 31092412 - 31202056 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030138.2 NP_084414.1 19 - 20 2175 66
GenBank mRNA AB490692.1 - 20 - 21 1949 66
AK029343.1 - 19 - 20 2099 66
AK129041.1 - 19 - 20 2152 66
AK147319.1 - 18 - 19 2047 66
BC046455.1 - 6 - 7 718 66

View Full Product Details