The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038018.4 | NP_001033107.1 | 4 - 5 | 524 | 68 |
NM_001112711.2 | NP_001106182.1 | 4 - 5 | 524 | 68 | |
NM_001286063.1 | NP_001272992.1 | 4 - 5 | 504 | 68 | |
NM_001286064.1 | NP_001272993.1 | 4 - 5 | 524 | 68 | |
NM_001286065.1 | NP_001272994.1 | 4 - 5 | 524 | 68 | |
NM_001286066.1 | NP_001272995.1 | 4 - 5 | 524 | 68 | |
NM_011938.4 | NP_036068.2 | 4 - 5 | 524 | 68 | |
GenBank mRNA | AF040747.1 | - | 4 - 5 | 394 | 68 |
AF040748.1 | - | 4 - 5 | 394 | 68 | |
AF040749.1 | - | 4 - 5 | 394 | 68 | |
AK051405.1 | - | 4 - 5 | 505 | 68 | |
AK146119.1 | - | 4 - 5 | 399 | 68 | |
AK161142.1 | - | 2 - 3 | 156 | 68 | |
AK179644.1 | - | 1 - 2 | 55 | 68 | |
BC057206.1 | - | 4 - 5 | 513 | 68 | |
BC075719.1 | - | 4 - 5 | 405 | 68 | |
Y15797.1 | - | 4 - 5 | 346 | 68 | |
Y15798.1 | - | 4 - 5 | 368 | 68 | |
Y15799.1 | - | 4 - 5 | 351 | 68 | |
Y15800.1 | - | 4 - 5 | 346 | 68 |