Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Usp11
Entrez Gene ID
236733
Gene Name
ubiquitin specific peptidase 11
Gene Aliases
6230415D12Rik, mKIAA4085
Chromosome Location
Chr.X: 20703909 - 20720539 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145628.4 NP_663603.3 6 - 7 773 74
GenBank mRNA AK031767.1 - 6 - 7 752 74
AK049053.1 - 6 - 7 1187 74
AK166523.1 - 6 - 7 749 74
AK220478.1 - 5 - 6 486 74
BC005470.1 - 4 - 5 342 74

View Full Product Details