The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167775.1 | NP_001161247.1 | 3 - 4 | 904 | 94 |
NM_145627.2 | NP_663602.1 | 3 - 4 | 617 | 94 | |
GenBank mRNA | AK145991.1 | - | 3 - 4 | 617 | 94 |
AK153736.1 | - | 3 - 4 | 905 | 94 | |
BC004674.1 | - | 2 - 3 | 302 | 94 |