The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113325.2 | NP_001106796.1 | 3 - 4 | 872 | 87 |
NM_001252403.1 | NP_001239332.1 | 3 - 4 | 499 | 87 | |
NM_008165.4 | NP_032191.2 | 3 - 4 | 872 | 87 | |
GenBank mRNA | AB102773.1 | - | 3 - 4 | 456 | 87 |
AB102774.1 | - | 3 - 4 | 456 | 87 | |
AK049958.1 | - | 3 - 4 | 682 | 87 | |
AK135107.1 | - | 3 - 4 | 500 | 87 | |
BC056397.1 | - | 3 - 4 | 534 | 87 | |
BC060702.1 | - | 3 - 4 | 455 | 87 | |
JN180918.1 | - | - | 523 | 87 | |
JN180919.1 | - | 3 - 4 | 454 | 87 | |
JN180920.1 | - | 3 - 4 | 344 | 87 | |
X57497.1 | - | 3 - 4 | 719 | 87 |