Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prkdc
Entrez Gene ID
19090
Gene Name
protein kinase, DNA activated, catalytic polypeptide
Gene Aliases
AI326420, AU019811, DNA-PKcs, DNAPDcs, DNAPK, DNPK1, DOXNPH, HYRC1, XRCC7, dxnph, p460, scid, slip
Chromosome Location
Chr.16: 15637444 - 15842252 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011159.2 NP_035289.2 3 - 4 349 147
GenBank mRNA AB007544.1 - 3 - 4 326 147
AB011543.1 - 3 - 4 338 147
AK084827.1 - 3 - 4 350 147
D87521.1 - 3 - 4 326 147

View Full Product Details