Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prkdc
Entrez Gene ID
19090
Gene Name
protein kinase, DNA activated, catalytic polypeptide
Gene Aliases
AI326420, AU019811, DNA-PKcs, DNAPDcs, DNAPK, DNPK1, DOXNPH, HYRC1, XRCC7, dxnph, p460, scid, slip
Chromosome Location
Chr.16: 15637444 - 15842252 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011159.2 NP_035289.2 82 - 83 11678 99
GenBank mRNA AB007544.1 - 82 - 83 11655 99
AB011543.1 - 82 - 83 11667 99
AK088981.1 - 7 - 8 805 99
AK137732.1 - 9 - 10 1112 99
D83786.1 - 7 - 8 813 99
D87521.1 - 82 - 83 11655 99
U78157.1 - 2 - 3 141 99
U78158.1 - 2 - 3 141 99

View Full Product Details