Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phldb2
Entrez Gene ID
208177
Gene Name
pleckstrin homology-like domain, family B, member 2
Gene Aliases
AV253284, C820004H04Rik, LL5b, LL5beta
Chromosome Location
Chr.16: 45746230 - 45953701 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252442.1 NP_001239371.1 17 - 18 3864 131
NM_153412.4 NP_700461.2 16 - 17 3705 131
GenBank mRNA AF506821.1 - 16 - 17 3706 131
AK035655.1 - 11 - 12 1468 131
AK087133.1 - 1 - 2 93 131
BC050915.1 - 17 - 18 3720 131
BC060683.1 - 17 - 18 3720 131

View Full Product Details