Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map3k13
Entrez Gene ID
71751
Gene Name
mitogen-activated protein kinase kinase kinase 13
Gene Aliases
C130026N12Rik, LZK
Chromosome Location
Chr.16: 21825171 - 21933439 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172821.3 NP_766409.2 9 - 10 1641 114
GenBank mRNA DQ480427.1 - 9 - 10 1641 114

View Full Product Details