The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199308.2 | NP_955012.2 | 24 - 25 | 3335 | 67 |
GenBank mRNA | AK172971.1 | - | 4 - 5 | 547 | 67 |
BC024265.1 | - | 6 - 7 | 983 | 67 |