The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173069.3 | NP_775092.3 | 1 - 2 | 154 | 59 |
GenBank mRNA | BB610282.1 | - | 1 - 2 | 156 | 59 |