The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011173.2 | NP_035303.1 | 7 - 8 | 824 | 83 |
GenBank mRNA | AK145501.1 | - | 7 - 8 | 824 | 83 |
AK163032.1 | - | 7 - 8 | 852 | 83 | |
BC128316.1 | - | 7 - 8 | 786 | 83 | |
L27439.1 | - | 6 - 7 | 635 | 83 | |
Z25469.1 | - | 7 - 8 | 790 | 83 |