Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sacm1l
Entrez Gene ID
83493
Gene Name
SAC1 (suppressor of actin mutations 1, homolog)-like (S. cerevisiae)
Gene Aliases
SAC1, Sac1p, mKIAA0851
Chromosome Location
Chr.9: 123529882 - 123592598 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030692.4 NP_109617.1 19 - 20 1639 72
GenBank mRNA AJ245720.1 - 19 - 20 1769 72
AJ297526.1 - 19 - 20 1640 72
AK089174.1 - 19 - 20 1723 72
AK122390.1 - 19 - 20 1704 72
AK154759.1 - 19 - 20 1739 72
AK161111.1 - 19 - 20 1673 72
AK171080.1 - 19 - 20 1723 72
BC005489.1 - 10 - 11 848 72
BC117752.1 - 19 - 20 1640 72
BC117753.1 - 19 - 20 1640 72

View Full Product Details