Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmprss15
Entrez Gene ID
19146
Gene Name
transmembrane protease, serine 15
Gene Aliases
Entk, Prss7
Chromosome Location
Chr.16: 78953008 - 79091097 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008941.3 NP_032967.1 19 - 20 2237 78
NM_178855.4 NP_849186.2 18 - 19 2192 78
GenBank mRNA AK033582.1 - 18 - 19 2194 78
AK038356.1 - 10 - 11 1186 78
BC117917.1 - 19 - 20 2236 78
BC117918.1 - 19 - 20 2236 78
U73378.1 - 19 - 20 2232 78

View Full Product Details