Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chodl
Entrez Gene ID
246048
Gene Name
chondrolectin
Gene Aliases
3110074E07Rik, MT75, PRED12
Chromosome Location
Chr.16: 78930864 - 78951733 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139134.3 NP_624360.2 2 - 3 811 64
GenBank mRNA AF311699.1 - 2 - 3 634 64
AK014255.1 - 2 - 3 522 64
AK031063.1 - 2 - 3 812 64
AK076523.1 - 2 - 3 811 64
AK140910.1 - 2 - 3 812 64
BC117071.1 - 2 - 3 482 64
BC117073.1 - 2 - 3 482 64
BC144738.1 - 2 - 3 482 64

View Full Product Details