Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Samsn1
Entrez Gene ID
67742
Gene Name
SAM domain, SH3 domain and nuclear localization signals, 1
Gene Aliases
4930571B16Rik, 930571B16Rik, Hacs1, SLy2
Chromosome Location
Chr.16: 75858794 - 75909266 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK016072.1 - - 868 71

View Full Product Details