The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013547.3 | NP_038575.2 | 6 - 7 | 603 | 78 |
NR_027778.1 | 5 - 6 | 514 | 78 | ||
GenBank mRNA | BC024369.1 | - | 5 - 6 | 417 | 78 |
BC044758.2 | - | 5 - 6 | 403 | 78 | |
BC055029.1 | - | 6 - 7 | 484 | 78 |