Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo40
Entrez Gene ID
207215
Gene Name
F-box protein 40
Gene Aliases
9830003A13Rik, mKIAA1195
Chromosome Location
Chr.16: 36963461 - 36990535 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK036385.1 - 1 - 2 113 70
AK036684.1 - 1 - 2 112 70

View Full Product Details