The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159419.1 | NP_001152891.1 | 25 - 26 | 2744 | 64 |
NM_198034.3 | NP_932151.2 | 24 - 25 | 2729 | 64 | |
GenBank mRNA | AK034344.1 | - | 10 - 11 | 894 | 64 |
AK046836.1 | - | 25 - 26 | 3104 | 64 | |
AK171920.1 | - | 24 - 25 | 2882 | 64 | |
AY562213.1 | - | 14 - 15 | 1288 | 65 | |
AY562214.1 | - | 14 - 15 | 1288 | 64 | |
BC025888.1 | - | 14 - 15 | 1379 | 64 | |
BC079584.1 | - | 24 - 25 | 2729 | 64 |