Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Retnlb
Entrez Gene ID
57263
Gene Name
resistin like beta
Gene Aliases
9030012B21Rik, Fizz2, RELMbeta, Relmb, Xcp3
Chromosome Location
Chr.16: 48816856 - 48818892 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023881.4 NP_076370.3 1 - 2 215 122
GenBank mRNA AK020240.1 - 1 - 2 103 122

View Full Product Details