The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028680.3 | NP_082956.2 | 10 - 11 | 1156 | 81 |
GenBank mRNA | AF274590.1 | - | 10 - 11 | 1174 | 81 |
AK040328.1 | - | 7 - 8 | 727 | 81 | |
AK047217.1 | - | 10 - 11 | 1156 | 81 | |
BC138143.1 | - | 10 - 11 | 1130 | 81 | |
BC138144.1 | - | 10 - 11 | 1130 | 81 | |
DQ266091.1 | - | 10 - 11 | 1197 | 81 |