The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025839.3 | NP_080115.1 | 1 - 2 | 183 | 58 |
GenBank mRNA | AF435792.1 | - | 1 - 2 | 195 | 58 |
AK002378.1 | - | 1 - 2 | 176 | 58 | |
AK003241.1 | - | 1 - 2 | 184 | 58 | |
AK007897.1 | - | 1 - 2 | 179 | 58 | |
AK019936.1 | - | 1 - 2 | 186 | 58 | |
AK159841.1 | - | 1 - 2 | 176 | 58 | |
BC025569.1 | - | 1 - 2 | 182 | 58 | |
BC028494.1 | - | 1 - 2 | 176 | 58 |