Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alg1
Entrez Gene ID
208211
Gene Name
asparagine-linked glycosylation 1 (beta-1,4-mannosyltransferase)
Gene Aliases
HMAT1, HMT1
Chromosome Location
Chr.16: 5233621 - 5244907 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145362.2 NP_663337.2 3 - 4 419 64
GenBank mRNA AK145871.1 - 3 - 4 419 64
BC011281.1 - 2 - 3 326 64

View Full Product Details