Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubn1
Entrez Gene ID
170644
Gene Name
ubinuclein 1
Gene Aliases
1110029L11Rik, 2610108L02Rik, AA673476, AW124741
Chromosome Location
Chr.16: 5050068 - 5086289 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026666.3 NP_080942.1 18 - 19 4227 60
GenBank mRNA AK084046.1 - 4 - 5 1589 60
AK147571.1 - 18 - 19 4228 60
BC098372.1 - 16 - 17 3387 60

View Full Product Details