Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bbx
Entrez Gene ID
70508
Gene Name
bobby sox homolog (Drosophila)
Gene Aliases
5530401J07Rik, 5730403O13Rik
Chromosome Location
Chr.16: 50191844 - 50432461 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027444.3 NP_081720.2 12 - 13 2344 101
GenBank mRNA AF454943.1 - 12 - 13 2337 101
AF454944.1 - 10 - 11 2209 101
AK029532.1 - 10 - 11 2211 101
AK029747.1 - 13 - 14 2458 101
AK049516.1 - 12 - 13 2330 101
AK167724.1 - 12 - 13 2475 101
BC066821.1 - 10 - 11 1971 101

View Full Product Details