Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bbx
Entrez Gene ID
70508
Gene Name
bobby sox homolog (Drosophila)
Gene Aliases
5530401J07Rik, 5730403O13Rik
Chromosome Location
Chr.16: 50191844 - 50432461 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027444.3 NP_081720.2 14 - 15 2559 89
GenBank mRNA AF454943.1 - 14 - 15 2552 89
AF454944.1 - 12 - 13 2424 89
AK041632.1 - 2 - 3 187 90
AK211657.1 - 2 - 3 142 89
BC066821.1 - 13 - 14 2276 89

View Full Product Details