The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081190.1 | NP_001074659.1 | 7 - 8 | 897 | 67 |
GenBank mRNA | AK141642.1 | - | 3 - 4 | 322 | 67 |
BC151105.1 | - | 7 - 8 | 947 | 67 |