The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024273.2 | NP_077235.2 | 2 - 3 | 489 | 78 |
GenBank mRNA | AK012774.1 | - | 2 - 3 | 463 | 78 |
AK081781.1 | - | 2 - 3 | 432 | 78 | |
BC002243.1 | - | 2 - 3 | 451 | 78 | |
CJ056343.1 | - | 2 - 3 | 491 | 78 |