The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019827.6 | NP_062801.1 | 7 - 8 | 2342 | 112 |
GenBank mRNA | AB066114.1 | - | 8 - 9 | 2341 | 112 |
AF156099.2 | - | 7 - 8 | 983 | 112 | |
AK154293.1 | - | 8 - 9 | 1820 | 112 | |
AK164048.1 | - | 6 - 7 | 694 | 112 | |
AK170965.1 | - | 8 - 9 | 1789 | 112 | |
BC006936.1 | - | 7 - 8 | 849 | 112 | |
BC060743.1 | - | 8 - 9 | 2269 | 112 |