The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300827.1 | NP_001287756.1 | 7 - 8 | 1036 | 136 |
NM_172380.4 | NP_759012.1 | 7 - 8 | 833 | 136 | |
GenBank mRNA | AK031608.1 | - | 7 - 8 | 772 | 136 |
AK035948.1 | - | 7 - 8 | 781 | 136 | |
AK036224.1 | - | 7 - 8 | 763 | 136 | |
AK041321.1 | - | 7 - 8 | 771 | 136 | |
AK045278.1 | - | 7 - 8 | 981 | 136 | |
AK079886.1 | - | 7 - 8 | 803 | 137 | |
AK167641.1 | - | 7 - 8 | 1003 | 136 | |
BC026809.1 | - | 7 - 8 | 784 | 136 |