Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd80
Entrez Gene ID
12519
Gene Name
CD80 antigen
Gene Aliases
B71, Cd28l, Ly-53, Ly53, MIC17, TSA1
Chromosome Location
Chr.16: 38455568 - 38498546 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009855.2 NP_033985.3 3 - 4 964 80
GenBank mRNA AF065893.1 - 3 - 4 773 80
AF065894.1 - 3 - 4 773 80
AF065895.1 - 3 - 4 773 80
AF065896.1 - 3 - 4 773 80
AJ278965.1 - 3 - 4 716 80
AK154329.1 - 4 - 5 1073 80
AY278186.1 - 3 - 4 764 80
BC131959.1 - 3 - 4 784 80
BC145843.1 - 3 - 4 784 80
X60958.1 - 3 - 4 964 80

View Full Product Details