Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scaf4
Entrez Gene ID
224432
Gene Name
SR-related CTD-associated factor 4
Gene Aliases
AA517739, Sfrs15, Sra4, Srsf15
Chromosome Location
Chr.16: 90228951 - 90284498 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178923.3 NP_849254.2 5 - 6 844 93
GenBank mRNA AK045266.1 - 5 - 6 841 93
AK173120.1 - 5 - 6 922 93
AK216612.1 - 2 - 3 190 93
BC023734.1 - 2 - 3 245 93
BC041782.1 - 2 - 3 216 93
BC053096.1 - 5 - 6 739 93
BC057592.1 - 5 - 6 844 93

View Full Product Details