The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028420.2 | NP_082696.1 | 1 - 2 | 259 | 98 |
GenBank mRNA | AK013788.1 | - | 1 - 2 | 206 | 98 |
BC147550.1 | - | 1 - 2 | 236 | 98 | |
BC147736.2 | - | 1 - 2 | 236 | 98 | |
BC147745.1 | - | 1 - 2 | 236 | 98 | |
BC157896.1 | - | 1 - 2 | 236 | 98 | |
CX215091.1 | - | 1 - 2 | 258 | 97 |