Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abcc1
Entrez Gene ID
17250
Gene Name
ATP-binding cassette, sub-family C (CFTR/MRP), member 1
Gene Aliases
Abcc1a, Abcc1b, MRP, Mdrap, Mrp1
Chromosome Location
Chr.16: 14361521 - 14474879 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008576.3 NP_032602.1 24 - 25 3621 65
GenBank mRNA AK029876.1 - 24 - 25 3622 65
AK049174.1 - 8 - 9 1388 65
AK140421.1 - 2 - 3 398 65
BC090617.1 - 24 - 25 3592 65

View Full Product Details