Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myh11
Entrez Gene ID
17880
Gene Name
myosin, heavy polypeptide 11, smooth muscle
Gene Aliases
AV071570, SM1, SM2, smMHC
Chromosome Location
Chr.16: 14194527 - 14291408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161775.1 NP_001155247.1 27 - 28 3799 63
NM_013607.2 NP_038635.2 27 - 28 3799 63
GenBank mRNA AK137226.1 - 28 - 29 3776 63
AK173045.1 - 27 - 28 3763 63
BC026142.1 - 27 - 28 3784 63
D85923.1 - 27 - 28 3754 63
D85924.1 - 27 - 28 3754 63

View Full Product Details