Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdxdc1
Entrez Gene ID
94184
Gene Name
pyridoxal-dependent decarboxylase domain containing 1
Gene Aliases
2210010A19Rik, AA415817, Kiaa0251-hp
Chromosome Location
Chr.16: 13833161 - 13903145 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039533.2 NP_001034622.1 12 - 13 1301 80
NM_001291017.1 NP_001277946.1 12 - 13 1301 80
NM_053181.3 NP_444411.2 12 - 13 1301 80
GenBank mRNA AK004611.1 - 12 - 13 1178 80
AK030613.1 - 12 - 13 1132 80
AK033107.1 - 12 - 13 1132 80
AK054329.1 - 12 - 13 1284 80
AK075829.1 - 5 - 6 424 80
AK163696.1 - 12 - 13 1172 80
AK170661.1 - 12 - 13 1288 80

View Full Product Details