Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfrc
Entrez Gene ID
22042
Gene Name
transferrin receptor
Gene Aliases
2610028K12Rik, AI195355, AI426448, AU015758, CD71, E430033M20Rik, Mtvr-1, Mtvr1, TFR, TFR1, TR, Trfr, p90
Chromosome Location
Chr.16: 32608896 - 32632794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011638.4 NP_035768.1 17 - 18 2078 137
GenBank mRNA AK088961.1 - 17 - 18 1992 137
BC054522.1 - 17 - 18 1963 137
BC067403.1 - 3 - 4 694 137
X57349.1 - 16 - 17 1914 137

View Full Product Details