The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163159.1 | NP_001156631.1 | 7 - 8 | 997 | 126 |
NM_001163160.1 | NP_001156632.1 | 7 - 8 | 931 | 126 | |
NM_009981.4 | NP_034111.1 | 7 - 8 | 833 | 126 | |
GenBank mRNA | AK076050.1 | - | 7 - 8 | 758 | 126 |
AK076830.1 | - | 7 - 8 | 932 | 126 | |
BC018313.1 | - | 6 - 7 | 716 | 126 | |
L28956.1 | - | 7 - 8 | 802 | 126 | |
Z12302.1 | - | 7 - 8 | 792 | 126 |