The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144854.2 | NP_659103.1 | 1 - 2 | 266 | 82 |
GenBank mRNA | AK041664.1 | - | 1 - 2 | 176 | 82 |
AY033899.1 | - | 1 - 2 | 281 | 82 | |
BC115624.1 | - | 1 - 2 | 40 | 82 | |
BC115625.1 | - | 1 - 2 | 40 | 82 | |
BX519631.1 | - | 1 - 2 | 267 | 82 |