The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081255.1 | NP_001074724.1 | 20 - 21 | 2230 | 77 |
GenBank mRNA | AK134026.1 | - | 10 - 11 | 862 | 77 |
AK155550.1 | - | 20 - 21 | 2228 | 77 | |
BC151058.1 | - | 20 - 21 | 2208 | 77 | |
BC157924.1 | - | 19 - 20 | 2100 | 77 | |
BC172144.1 | - | 18 - 19 | 1950 | 77 |