The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013371.2 | NP_001013389.2 | 1 - 2 | 336 | 111 |
GenBank mRNA | AK090152.1 | - | 1 - 2 | 226 | 111 |
AK146802.1 | - | 1 - 2 | 244 | 111 | |
BC120494.1 | - | 1 - 2 | 277 | 111 | |
BC137694.1 | - | 1 - 2 | 277 | 111 | |
CN668148.1 | - | 1 - 2 | 336 | 111 |