The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013371.2 | NP_001013389.2 | 2 - 3 | 582 | 62 |
GenBank mRNA | AK090152.1 | - | 2 - 3 | 472 | 62 |
AK146802.1 | - | 2 - 3 | 490 | 62 | |
BC085093.1 | - | 2 - 3 | 487 | 62 | |
BC120494.1 | - | 2 - 3 | 523 | 62 | |
BC137694.1 | - | 2 - 3 | 523 | 62 | |
CN668148.1 | - | 2 - 3 | 581 | 62 |