The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173026.2 | NP_766614.2 | 10 - 11 | 2889 | 68 |
GenBank mRNA | BC055403.1 | - | 2 - 3 | 1049 | 68 |