Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb11
Entrez Gene ID
271377
Gene Name
zinc finger and BTB domain containing 11
Gene Aliases
9230110G02Rik, ZNF-U69274
Chromosome Location
Chr.16: 55972697 - 56008913 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173026.2 NP_766614.2 7 - 8 2435 92
GenBank mRNA AK033794.1 - 4 - 5 1116 92

View Full Product Details