Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep97
Entrez Gene ID
74201
Gene Name
centrosomal protein 97
Gene Aliases
2810403B08Rik, 4932439K18Rik, AA437768, E130116N02Rik, Lrriq2
Chromosome Location
Chr.16: 55899888 - 55934848 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159364.1 NP_001152836.1 3 - 4 398 69
NM_001159365.1 NP_001152837.1 3 - 4 398 69
NM_001159366.1 NP_001152838.1 3 - 4 398 69
NM_028815.4 NP_083091.1 3 - 4 398 69
GenBank mRNA AK012971.2 - 3 - 4 383 69
AK040079.1 - 3 - 4 405 69
AK046486.1 - 3 - 4 947 69
AK079884.1 - 3 - 4 366 69
AK080249.1 - 3 - 4 384 69
AK154885.1 - 3 - 4 377 69
BC065773.1 - 3 - 4 347 69
BC067049.1 - 3 - 4 398 69

View Full Product Details