Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gc
Entrez Gene ID
14473
Gene Name
group specific component
Gene Aliases
DBP, VDB
Chromosome Location
Chr.5: 89417511 - 89457898 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008096.2 NP_032122.1 5 - 6 782 73
GenBank mRNA AK010965.1 - 5 - 6 606 73
BC010762.1 - 5 - 6 645 73
BC051395.1 - 5 - 6 649 73
M55413.1 - 5 - 6 592 73

View Full Product Details