The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007764.5 | NP_031790.2 | 1 - 2 | 808 | 76 |
GenBank mRNA | AK048641.1 | - | 1 - 2 | 806 | 76 |
AK052315.1 | - | 1 - 2 | 808 | 76 | |
AK163981.1 | - | 1 - 2 | 780 | 76 | |
BC131984.1 | - | 1 - 2 | 575 | 76 | |
BC131986.1 | - | 1 - 2 | 575 | 76 | |
X90648.1 | - | 1 - 2 | 647 | 76 |