Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc74a
Entrez Gene ID
72315
Gene Name
coiled-coil domain containing 74A
Gene Aliases
2310015A05Rik
Chromosome Location
Chr.16: 17645903 - 17650739 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166164.1 NP_001159636.1 2 - 3 364 96
GenBank mRNA AK009347.1 - 2 - 3 217 96

View Full Product Details