Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frmpd1
Entrez Gene ID
666060
Gene Name
FERM and PDZ domain containing 1
Gene Aliases
BC031840, mKIAA0967
Chromosome Location
Chr.4: 45184417 - 45285936 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081172.2 NP_001074641.1 5 - 6 469 108
GenBank mRNA AK122409.1 - 2 - 3 95 108
BB660211.1 - 5 - 6 470 108
BC158060.1 - 5 - 6 433 108

View Full Product Details