Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmod1
Entrez Gene ID
21916
Gene Name
tropomodulin 1
Gene Aliases
E-Tmod, Tmod
Chromosome Location
Chr.4: 46038940 - 46116032 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021883.2 NP_068683.1 3 - 4 413 75
GenBank mRNA AK132878.1 - 2 - 3 246 75
BC106849.1 - 2 - 3 301 75
BC106850.1 - 2 - 3 301 75
S76831.1 - 2 - 3 340 75

View Full Product Details